5-Formyl-2-(morpholin-4-yl)benzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Formyl-2-morpholinobenzonitrile

059582-500MG 059582-1G 059582-5G
101918-540EA 193.85 USD
101918-540 101918-542 101918-544
5-Formyl-2-(morpholin-4-yl)benzonitrile ≥95%
5-Formyl-2-(morpholin-4-yl)benzonitrile

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD18651781
CAS Number: 1272756-61-2

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR