2-Fluoro-5-(hydroxymethyl)benzonitrile ≥95%

Supplier: Matrix Scientific

059581-500MG 059581-1G 059581-5G
101918-534EA 49.23 USD
101918-534 101918-536 101918-538
2-Fluoro-5-(hydroxymethyl)benzonitrile ≥95%
2-Fluoro-5-(hydroxymethyl)benzonitrile

Hazard: Irritant

Formula: C₈H₆FNO
Melting Pt: 45…47 °C
MDL Number: MFCD03094340
CAS Number: 227609-85-0

Order Now

Specification Test Results

Melting point 45-47°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR