4-Fluoro-3-(furan-2-yl)-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

059580-500MG 059580-1G
101918-530EA 600 USD
101918-530 101918-532
4-Fluoro-3-(furan-2-yl)-1H-pyrazol-5-amine ≥95%
4-Fluoro-3-(furan-2-yl)-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₇H₆FN₃O
Melting Pt: 137…139 °C
MDL Number: MFCD18651765
CAS Number: 1323438-99-8

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR