6-Ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Ethylpyrimidine-2,4(1H,3H)-dione

059579-500MG 059579-1G
101918-526EA 600 USD
101918-526 101918-528
6-Ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione ≥95%
6-Ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD18377697
CAS Number: 15043-03-5

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR