Ethyl-4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

059577-500MG 059577-1G 059577-5G
101918-514EA 193.85 USD
101918-514 101918-516 101918-518
Ethyl-4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate hydrochloride ≥95%
Ethyl-4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O₂S
Melting Pt: 220…222 °C
MDL Number: MFCD18380662
CAS Number: 1269152-25-1

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR