Ethyl-3-iodo-5-nitro-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

059576-500MG 059576-1G
101918-510EA 600 USD
101918-510 101918-512
Ethyl-3-iodo-5-nitro-1H-indole-2-carboxylate ≥95%
Ethyl-3-iodo-5-nitro-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₉IN₂O₄
Melting Pt: 276…277 °C
MDL Number: MFCD18651761
CAS Number: 494854-46-5

Order Now

Specification Test Results

Melting point 276-277°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR