Ethyl (2Z)-2-cyano-3-hydroxy-3-(4-methylphenyl)prop-2-enoate ≥95%

Supplier: Matrix Scientific

059571-500MG 059571-1G 059571-5G
101918-486EA 193.85 USD
101918-486 101918-488 101918-490
Ethyl (2Z)-2-cyano-3-hydroxy-3-(4-methylphenyl)prop-2-enoate ≥95%
Ethyl (2Z)-2-cyano-3-hydroxy-3-(4-methylphenyl)prop-2-enoate

Hazard: Irritant

Formula: C₁₃H₁₃NO₃
Melting Pt: 43…45 °C
MDL Number: MFCD18651762
CAS Number: 91024-54-3

Order Now

Specification Test Results

Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR