Ethyl-1-(2-cyano-4-formylphenyl)piperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

059570-500MG 059570-1G 059570-5G
101918-480EA 193.85 USD
101918-480 101918-482 101918-484
Ethyl-1-(2-cyano-4-formylphenyl)piperidine-4-carboxylate ≥95%
Ethyl-1-(2-cyano-4-formylphenyl)piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₃
Melting Pt: 76…78 °C
MDL Number: MFCD18651778
CAS Number: 1272756-67-8

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR