Ethyl-2-(2-carbamimidamido-1,3-thiazol-4-yl)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

059567-500MG 059567-1G 059567-5G
101918-466EA 193.85 USD
101918-466 101918-468 101918-470
Ethyl-2-(2-carbamimidamido-1,3-thiazol-4-yl)acetate hydrochloride ≥95%
Ethyl-2-(2-carbamimidamido-1,3-thiazol-4-yl)acetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₄O₂S
Melting Pt: 133…135 °C
MDL Number: MFCD18651780
CAS Number: 1272756-56-5

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR