Ethyl-2-carbamimidamido-1,3-thiazole-4-carboxylate hydrobromide ≥95%

Supplier: Matrix Scientific

059565-500MG 059565-1G 059565-5G
101918-454EA 193.85 USD
101918-454 101918-456 101918-458
Ethyl-2-carbamimidamido-1,3-thiazole-4-carboxylate hydrobromide ≥95%
Ethyl-2-carbamimidamido-1,3-thiazole-4-carboxylate hydrobromide

Hazard: Irritant

Formula: C₇H₁₁BrN₄O₂S
Melting Pt: 188…190 °C
MDL Number: MFCD18651775
CAS Number: 231621-74-2

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR