Ethyl-3-bromo-5-nitro-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

059564-500MG 059564-1G
101918-450EA 600 USD
101918-450 101918-452
Ethyl-3-bromo-5-nitro-1H-indole-2-carboxylate ≥95%
Ethyl-3-bromo-5-nitro-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₉BrN₂O₄
Melting Pt: 258…259 °C
MDL Number: MFCD18651771
CAS Number: 183384-45-4

Order Now

Specification Test Results

Melting point 258-259°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR