Ethyl-1-[(2-aminophenyl)carbonyl]piperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

059563-500MG 059563-1G
101918-446EA 403.08 USD
101918-446 101918-448
Ethyl-1-[(2-aminophenyl)carbonyl]piperidine-4-carboxylate ≥95%
Ethyl-1-[(2-aminophenyl)carbonyl]piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₃
Melting Pt: 103…104 °C
MDL Number: MFCD05258487
CAS Number: 696654-85-0

Order Now

Specification Test Results

Melting point 103-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR