1-(4,5-Dimethyl-1,3-thiazol-2-yl)guanidinehydrobromide ≥95%

Supplier: Matrix Scientific

059560-500MG 059560-1G 059560-5G
101918-434EA 193.85 USD
101918-434 101918-436 101918-438
1-(4,5-Dimethyl-1,3-thiazol-2-yl)guanidinehydrobromide ≥95%
1-(4,5-Dimethyl-1,3-thiazol-2-yl)guanidinehydrobromide

Hazard: Irritant

Formula: C₆H₁₁BrN₄S
Melting Pt: 279…281 °C
MDL Number: MFCD18651774
CAS Number: 85207-84-7

Order Now

Specification Test Results

Melting point 279-281°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR