1-(2-Chloropyridin-3-yl)-3-(dimethylamino)prop-2-en-1-one ≥95%

Supplier: Matrix Scientific

059555-500MG 059555-1G 059555-5G
101918-414EA 600 USD
101918-414 101918-416 101918-418
1-(2-Chloropyridin-3-yl)-3-(dimethylamino)prop-2-en-1-one ≥95%
1-(2-Chloropyridin-3-yl)-3-(dimethylamino)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O
Melting Pt: 73…76 °C
MDL Number: MFCD18651787
CAS Number: 166196-84-5

Order Now

Specification Test Results

Melting point 73-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR