6-Bromo-4-oxo-1,4-dihydroquinoline-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

059549-500MG 059549-1G
101918-396EA 600 USD
101918-396 101918-398
6-Bromo-4-oxo-1,4-dihydroquinoline-8-carboxylic acid ≥95%
6-Bromo-4-oxo-1,4-dihydroquinoline-8-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrNO₃
MW: 268.07 g/mol
Melting Pt: >300 °C
MDL Number: MFCD18651770
CAS Number: 1272756-55-4

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR