2,3-Bis(5-amino-2-chlorophenyl)propanenitrile ≥95%

Supplier: Matrix Scientific

059544-500MG 059544-1G
101918-374EA 995.38 USD
101918-374 101918-376
2,3-Bis(5-amino-2-chlorophenyl)propanenitrile ≥95%
2,3-Bis(5-amino-2-chlorophenyl)propanenitrile

Hazard: Irritant

Formula: C₁₅H₁₃Cl₂N₃
Melting Pt: 134…137 °C
MDL Number: MFCD18651763
CAS Number: 1272756-66-7

Order Now

Specification Test Results

Melting point 134-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR