2-Amino-N-(2H-1,3-benzodioxol-5-yl)benzamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-amino-N-1,3-benzodioxol-5-ylbenzamide

059538-500MG 059538-1G
101918-354EA 221.54 USD
101918-354 101918-356
2-Amino-N-(2H-1,3-benzodioxol-5-yl)benzamide ≥95%
2-Amino-N-(2H-1,3-benzodioxol-5-yl)benzamide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₃
MW: 256.26 g/mol
Melting Pt: 113…114 °C
MDL Number: MFCD04606709
CAS Number: 571158-97-9

Order Now

Specification Test Results

Melting point 113-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR