Methyl-2-ethyl-5-methoxy-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054900-500MG 054900-1G
101908-986EA 556.92 USD
101908-986 101908-988
Methyl-2-ethyl-5-methoxy-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-2-ethyl-5-methoxy-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
Melting Pt: 84…86 °C
MDL Number: MFCD16140360
CAS Number: 1221792-19-3

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR