Methyl-2-ethyl-7-methoxy-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

054899-500MG 054899-1G
101908-982EA 556.92 USD
101908-982 101908-984
Methyl-2-ethyl-7-methoxy-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-2-ethyl-7-methoxy-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
Melting Pt: 104…106 °C
MDL Number: MFCD16140359
CAS Number: 1221792-17-1

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR