Methyl-2-cyano-8-methoxy-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

054898-500MG 054898-1G
101908-978EA 747.69 USD
101908-978 101908-980
Methyl-2-cyano-8-methoxy-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate ≥95%
Methyl-2-cyano-8-methoxy-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₄
Melting Pt: 163…165 °C
MDL Number: MFCD16140358
CAS Number: 1221792-69-3

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR