Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 4-chloro-3-methoxy-5-nitrobenzoate

054897-500MG 054897-1G 054897-5G
101908-972EA 458.46 USD
101908-972 101908-974 101908-976
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate ≥97%
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₉H₈ClNO₅
Melting Pt: 103…105 °C
MDL Number: MFCD16140357
CAS Number: 63603-09-8

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR