Methyl-2-(benzyloxy)-5-methoxy-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(benzyloxy)-5-methoxy-3-nitrobenzoate

054896-500MG 054896-1G 054896-5G
101908-966EA 278.46 USD
101908-966 101908-968 101908-970
Methyl-2-(benzyloxy)-5-methoxy-3-nitrobenzenecarboxylate ≥95%
Methyl-2-(benzyloxy)-5-methoxy-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₅NO₆
MW: 317.3 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD16140356
CAS Number: 1221792-98-8

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR