Methyl-3-amino-4-chloro-5-methoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-amino-4-chloro-5-methoxybenzoate
054895-500MG
054895-1G
101908-962EA
467.69
USD
101908-962
101908-964
Methyl-3-amino-4-chloro-5-methoxybenzenecarboxylate ≥95%
Methyl-3-amino-4-chloro-5-methoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀ClNO₃ Melting Pt: 93…95 °C |
MDL Number:
MFCD16140355 CAS Number: 63603-10-1 |
Specification Test Results
Melting point | 93-95°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...