Methyl-3-amino-2-(benzyloxy)-5-methoxybenzoate hydrochloride ≥95%

Supplier: Matrix Scientific

054894-500MG 054894-1G 054894-5G
101908-956EA 375.38 USD
101908-956 101908-958 101908-960
Methyl-3-amino-2-(benzyloxy)-5-methoxybenzoate hydrochloride ≥95%
Methyl-3-amino-2-(benzyloxy)-5-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈ClNO₄
MW: 323.78 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD16140354
CAS Number: 1221792-96-6

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR