Ethyl (E)-3-(4-pyridinyl)-2-propenoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(pyridin-4-yl)acrylate

054888-1G 054888-5G
101908-928EA 158.46 USD
101908-928 101908-930
Ethyl (E)-3-(4-pyridinyl)-2-propenoate ≥95%
Ethyl (E)-3-(4-pyridinyl)-2-propenoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD07778427
CAS Number: 24489-96-1

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR