Ethyl-3-cyano-6-cyclopropyl-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

054886-500MG 054886-1G
101908-918EA 747.69 USD
101908-918 101908-920
Ethyl-3-cyano-6-cyclopropyl-2-pyridinecarboxylate ≥95%
Ethyl-3-cyano-6-cyclopropyl-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 44…46 °C
MDL Number: MFCD16140350
CAS Number: 1221791-78-1

Order Now

Specification Test Results

Melting point 44-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR