Ethyl (1S,5R,6R)-3-benzyl-2,4-dioxo-3-azabicyclo-[3.1.0]hexane-6-carboxylate ≥95%

Supplier: Matrix Scientific

054885-500MG 054885-1G 054885-5G
101908-912EA 747.69 USD
101908-912 101908-914 101908-916
Ethyl (1S,5R,6R)-3-benzyl-2,4-dioxo-3-azabicyclo-[3.1.0]hexane-6-carboxylate ≥95%
Ethyl (1S,5R,6R)-3-benzyl-2,4-dioxo-3-azabicyclo-[3.1.0]hexane-6-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₅NO₄
MW: 273.29 g/mol
Melting Pt: 99…100 °C
MDL Number: MFCD16170316
CAS Number: 134575-06-7

Order Now

Specification Test Results

Melting point 99-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR