Ethyl-3-acetyl-5-chloro-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

054884-500MG 054884-1G
101908-908EA 692.31 USD
101908-908 101908-910
Ethyl-3-acetyl-5-chloro-2-pyridinecarboxylate ≥95%
Ethyl-3-acetyl-5-chloro-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₃
Melting Pt: 83…85 °C
MDL Number: MFCD16140348
CAS Number: 1221791-87-2

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR