1,9-Dithia-4,12-diazadispiro[4.2.4

Supplier: Matrix Scientific

054883-500MG 054883-1G
101908-904EA 278.46 USD
101908-904 101908-906
1,9-Dithia-4,12-diazadispiro[4.2.4
1,9-Dithia-4,12-diazadispiro[4.2.4^{8}.2^{5}]tetradecane dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀Cl₂N₂S₂
Melting Pt: 260…264 °C (dec.)
MDL Number: MFCD16140347
CAS Number: 1221792-22-8

Order Now

Specification Test Results

Melting point 260-264°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR