8-Cyano-8-phenyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

054879-500MG 054879-1G
101908-892EA 278.46 USD
101908-892 101908-894
8-Cyano-8-phenyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%
8-Cyano-8-phenyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₂S
Melting Pt: 203…205 °C (dec.)
MDL Number: MFCD16140343
CAS Number: 1236255-46-1

Order Now

Specification Test Results

Melting point 203-205°C(D

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR