4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}aniline ≥95%

Supplier: Matrix Scientific

054875-500MG 054875-1G
101908-878EA 375.38 USD
101908-878 101908-880
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}aniline ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}aniline

Hazard: Irritant

Formula: C₁₂H₈ClF₃N₂S
Melting Pt: 120…122 °C
MDL Number: MFCD13459218
CAS Number: 581792-83-8

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR