6-[(4-Chlorophenyl)sulfonyl]-3-pyridinylamine ≥95%

Supplier: Matrix Scientific

054873-500MG 054873-1G
101908-870EA 556.92 USD
101908-870 101908-872
6-[(4-Chlorophenyl)sulfonyl]-3-pyridinylamine ≥95%
6-[(4-Chlorophenyl)sulfonyl]-3-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₂S
Melting Pt: 199…201 °C
MDL Number: MFCD00084873
CAS Number: 52117-91-6

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR