5-(4-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione ≥95%

Supplier: Matrix Scientific

054869-500MG 054869-1G
101908-860EA 375.38 USD
101908-860 101908-862
5-(4-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione ≥95%
5-(4-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione

Hazard: Irritant

Formula: C₉H₈BrN₃S
MW: 270.16 g/mol
Melting Pt: 246…248 °C
MDL Number: MFCD03995808
CAS Number: 494194-50-2

Order Now

Specification Test Results

Melting point 246-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR