5-(3-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione ≥95%

Supplier: Matrix Scientific

054868-500MG 054868-1G
101908-856EA 375.38 USD
101908-856 101908-858
5-(3-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione ≥95%
5-(3-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione

Hazard: Irritant

Formula: C₉H₈BrN₃S
MW: 270.16 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD01816005
CAS Number: 333771-24-7

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR