3-Bromo-2,4-diiodo-5-methylthiophene ≥95%

Supplier: Matrix Scientific

054865-500MG 054865-1G 054865-5G
101908-842EA 181.54 USD
101908-842 101908-844 101908-846
3-Bromo-2,4-diiodo-5-methylthiophene ≥95%
3-Bromo-2,4-diiodo-5-methylthiophene

Hazard: Irritant

Formula: C₅H₃BrI₂S
Melting Pt: 75-76°C
MDL Number: MFCD16140335
CAS Number: 1221791-68-9

Order Now

Specification Test Results

Melting point 75-76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR