trans-1-Benzyl-4-(pyridin-2-yl)pyrrolidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

054864-500MG 054864-1G
101908-838EA 1109.23 USD
101908-838 101908-840
trans-1-Benzyl-4-(pyridin-2-yl)pyrrolidine-3-carboxylic acid ≥95%
trans-1-Benzyl-4-(pyridin-2-yl)pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₈N₂O₂
Melting Pt: 85…88 °C
MDL Number: MFCD16140334
CAS Number: 1221791-62-3

Order Now

Specification Test Results

Melting point 85-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR