2-(Benzyloxy)-5-methoxy-3-nitrobenzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
054863-500MG
054863-1G
054863-5G
101908-832EA
375.38
USD
101908-832
101908-834
101908-836
2-(Benzyloxy)-5-methoxy-3-nitrobenzenecarboxylic acid ≥95%
2-(Benzyloxy)-5-methoxy-3-nitrobenzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₃NO₆ Melting Pt: 134…135 °C |
MDL Number:
MFCD16140333 CAS Number: 1221791-72-5 |
Specification Test Results
Melting point | 134-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...