2-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetic acid ≥95%
Supplier: Matrix Scientific
054855-500MG
054855-1G
054855-5G
101908-802EA
356.92
USD
101908-802
101908-804
101908-806
2-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetic acid ≥95%
2-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetic acid
Hazard: Irritant
Formula:
C₁₀H₇F₃N₂O₂ Melting Pt: 182…183 °C |
MDL Number:
MFCD14546662 CAS Number: 1221792-70-6 |
Specification Test Results
Melting point | 182-183°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...