2-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetohydrazide ≥95%
Supplier: Matrix Scientific
054854-500MG
054854-1G
054854-5G
101908-796EA
375.38
USD
101908-796
101908-798
101908-800
2-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetohydrazide ≥95%
2-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetohydrazide
Hazard: Irritant
Formula:
C₁₀H₉F₃N₄O |
MDL Number:
MFCD16140326 CAS Number: 1221792-29-5 |
Specification Test Results
Melting point | 222°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...