1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride ≥95%

Supplier: Matrix Scientific

054848-500MG 054848-1G
101908-778EA 375.38 USD
101908-778 101908-780
1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride ≥95%
1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNOS
Melting Pt: 192…195 °C
MDL Number: MFCD16140323
CAS Number: 1221792-86-4

Order Now

Specification Test Results

Melting point 192-195°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR