tert-Butyl-3H-spiro[1,3-benzothiazole-2,4'-piperidine]-1'-carboxylate ≥95%
Supplier: Matrix Scientific
054847-500MG
054847-1G
101908-774EA
278.46
USD
101908-774
101908-776
tert-Butyl-3H-spiro[1,3-benzothiazole-2,4'-piperidine]-1'-carboxylate ≥95%
tert-Butyl-3H-spiro[1,3-benzothiazole-2,4'-piperidine]-1'-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₂N₂O₂S MW: 306.43 g/mol Melting Pt: 104…106 °C |
MDL Number:
MFCD16140322 CAS Number: 1221792-85-3 |
Specification Test Results
Melting point | 104-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...