tert-Butyl-2-oxo-1,4,8-triazaspiro[4.5]decane-8-carboxylate ≥95%

Supplier: Matrix Scientific

054846-500MG 054846-1G
101908-770EA 207.69 USD
101908-770 101908-772
tert-Butyl-2-oxo-1,4,8-triazaspiro[4.5]decane-8-carboxylate ≥95%
tert-Butyl-2-oxo-1,4,8-triazaspiro[4.5]decane-8-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁N₃O₃
MW: 255.32 g/mol
Melting Pt: 171…173 °C
MDL Number: MFCD16140321
CAS Number: 1221792-93-3

Order Now

Specification Test Results

Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR