1-(tert-Butyl)-5-methyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

054845-500MG 054845-1G 054845-5G
101908-764EA 341.54 USD
101908-764 101908-766 101908-768
1-(tert-Butyl)-5-methyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(tert-Butyl)-5-methyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Melting Pt: 132…133 °C
MDL Number: MFCD11810587
CAS Number: 288251-51-4

Order Now

Specification Test Results

Melting point 132-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR