tert-Butyl-4-(5-bromo-3-nitro-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
054842-500MG
054842-1G
054842-5G
101908-752EA
375.38
USD
101908-752
101908-754
101908-756
tert-Butyl-4-(5-bromo-3-nitro-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(5-bromo-3-nitro-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₉BrN₄O₄ Melting Pt: 100…101 °C |
MDL Number:
MFCD16140319 CAS Number: 1221792-73-9 |
Specification Test Results
Melting point | 100-101°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...