3H-Spiro[1,3-benzothiazole-2,4'-oxane] ≥95%

Supplier: Matrix Scientific

054835-500MG 054835-1G
101908-724EA 278.46 USD
101908-724 101908-726
3H-Spiro[1,3-benzothiazole-2,4'-oxane] ≥95%
3H-Spiro[1,3-benzothiazole-2,4'-oxane]

Hazard: Irritant

Formula: C₁₁H₁₃NOS
MW: 207.3 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD16140318
CAS Number: 77376-98-8

Order Now

Specification Test Results

Melting point 99-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR