8-Oxa-1-thia-4-azaspiro[4.5]decan-3-one ≥95%

Supplier: Matrix Scientific

054823-500MG 054823-1G
101908-676EA 278.46 USD
101908-676 101908-678
8-Oxa-1-thia-4-azaspiro[4.5]decan-3-one ≥95%
8-Oxa-1-thia-4-azaspiro[4.5]decan-3-one

Hazard: Irritant

Formula: C₇H₁₁NO₂S
Melting Pt: 130…132 °C
MDL Number: MFCD16140315
CAS Number: 1221792-94-4

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR