8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride ≥95%

Supplier: Matrix Scientific

054822-500MG 054822-1G
101908-672EA 278.46 USD
101908-672 101908-674
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride ≥95%
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNOS
Melting Pt: 166…167 °C
MDL Number: MFCD16140314
CAS Number: 1221791-86-1

Order Now

Specification Test Results

Melting point 166-167°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR