Octahydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine] ≥95%

Supplier: Matrix Scientific

054821-500MG 054821-1G
101908-668EA 375.38 USD
101908-668 101908-670
Octahydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine] ≥95%
Octahydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]

Hazard: Irritant

Formula: C₁₂H₂₁NS
Melting Pt: 41…43 °C
MDL Number: MFCD16140313
CAS Number: 1221792-92-2

Order Now

Specification Test Results

Melting point 41-43°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR