3',4',4'a,5',6',7',8',8'a-Octahydro-1'H,3H-spiro[1,3-benzothiazole-2,2'-naphthalene] ≥95%

Supplier: Matrix Scientific

054820-500MG 054820-1G
101908-664EA 278.46 USD
101908-664 101908-666
3',4',4'a,5',6',7',8',8'a-Octahydro-1'H,3H-spiro[1,3-benzothiazole-2,2'-naphthalene] ≥95%
3',4',4'a,5',6',7',8',8'a-Octahydro-1'H,3H-spiro[1,3-benzothiazole-2,2'-naphthalene]

Hazard: Irritant

Formula: C₁₆H₂₁NS
Melting Pt: 55…58 °C
MDL Number: MFCD16140312
CAS Number: 1221791-69-0

Order Now

Specification Test Results

Melting point 55-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR