Methyl-2-[6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate ≥95%

Supplier: Matrix Scientific

054814-500MG 054814-1G 054814-5G
101908-638EA 375.38 USD
101908-638 101908-640 101908-642
Methyl-2-[6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate ≥95%
Methyl-2-[6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate

Hazard: Irritant

Formula: C₁₁H₉F₃N₂O₂
Melting Pt: 89…90 °C
MDL Number: MFCD16140311
CAS Number: 1221792-68-2

Order Now

Specification Test Results

Melting point 89-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR